PYREX™ Disposable Round Bottom Threaded Culture Tubes with White Marking Spot, without Caps
PYREX™ Disposable Round Bottom Threaded Culture Tubes with White Marking Spot, without Caps
Actual product may vary
PYREX™ Disposable Round Bottom Threaded Culture Tubes with White Marking Spot, without Caps
For tissue culture and general bacteriological work
Have Questions?
Catalog NumberCapacity (Metric)Length (Metric)Diameter (Metric)Thread Size
FIS14-962A7.5 mL100 mm13 mm13-415
Catalog number FIS14-962A
Price (SGD)
2,041.00
CS
Add to cart
Capacity (Metric):
7.5 mL
Length (Metric):
100 mm
Diameter (Metric):
13 mm
Thread Size:
13-415
Price (SGD)
2,041.00
CS
Add to cart
Designed for both tissue culture and general bacteriological work. Tubes are made of borosilicate glass and are sold without caps.
For caps, see Cat. No. 99999.
Specifications
Design TypeThreaded
Capacity (Metric)7.5 mL
Length (Metric)100 mm
MaterialBorosilicate Glass
DisposableSingle-use
For Use WithFor tissue culture work and general bacteriological use
Label TypeWhite Marking Spot
PackagingBulk
Thread Size13-415
SterilityNon-sterile
GraduatedNo
Diameter (Metric)13 mm
Closure TypeNot Included
Product TypeCulture Tube
Unit SizeCS