Round-Bottom Polypropylene Test Tubes Without Cap
Round-Bottom Polypropylene Test Tubes Without Cap
Actual product may vary
Round-Bottom Polypropylene Test Tubes Without Cap
Manufactured from polypropylene for excellent chemical and thermal resistance. Falcon™Round-Bottom Polypropylene Tubes are tested to withstand 3000 x g centrifugation.
Have Questions?
Catalog NumberCapacity (Metric)Length (Metric)SterilityDiameter (Metric)
FIS14-959AA5 mL75 mmNon-sterile12 mm
Catalog number FIS14-959AA
Price (SGD)
302.00
CS
Add to cart
Capacity (Metric):
5 mL
Length (Metric):
75 mm
Sterility:
Non-sterile
Diameter (Metric):
12 mm
Price (SGD)
302.00
CS
Add to cart
  • Widely used and referenced in laboratory protocols
  • Sterile, nonpyrogenicity tested to less than 0.1 EU/mL (Cat. No. 352002 non-sterile)
The foundation for consistent research results
  • Provide reliable containment of laboratory fluid samples
  • Polypropylene tubes are best suited for applications requiring greater thermal and chemical stability
  • Medical-style packaging materials
  • 3,000 RCF rating (RCF claims refer to Relative Centrifugal Force measured in g-force for materials with a specific gravity of 1.0, used in an appropriate rotor with correct cushion and safety precautions. Tubes used with organic solvents at temperatures below 0°C may have lower RCF ratings.)
  • 17 x 100 mm tubes have blue printed graduations and white writing patch
  • Tube dimensions and volumes are approximate
Specifications
ColorClear
Description3000 x G
Capacity (Metric)5 mL
Length (Metric)75 mm
MaterialPolypropylene
DisposableSingle-use
Packaging1000/Cs.
SterilityNon-sterile
GraduatedNo
Diameter (Metric)12 mm
Closure TypeNot Included
Product TypeTest Tube
Unit SizeCS