PYREX™ Reusable Borosilicate Glass Tubes with Plain End
PYREX™ Reusable Borosilicate Glass Tubes with Plain End
Actual product may vary
PYREX™ Reusable Borosilicate Glass Tubes with Plain End
PYREX™ Reusable Borosilicate Glass Tubes with Plain End
PYREX™ Reusable Borosilicate Glass Tubes with Plain End
PYREX™ Reusable Borosilicate Glass Tubes with Plain End
PYREX™ Reusable Borosilicate Glass Tubes with Plain End
All sizes except 6, 22, and 25mm O.D. have marking spot
Have Questions?
Catalog NumberCapacity (Metric)Length (Metric)Diameter (Metric)
FIS14-957G20 mL150 mm16 mm
Catalog number FIS14-957G
Price (SGD)
235.00
PK
Add to cart
Capacity (Metric):
20 mL
Length (Metric):
150 mm
Diameter (Metric):
16 mm
Price (SGD)
235.00
PK
Add to cart
All sizes except 6, 22, and 25mm O.D. have marking spot
Rimless for greater convenience in plugging and rack storage. Walls and bottoms are of uniform thickness. Ends are fire-polished.
Cat. No 9820-12 is a Kahn tubes.
Cat. No. 9820-13 is a Wasserman tube.
Specifications
Design TypePlain Rimless
Capacity (Metric)20 mL
Length (Metric)150 mm
MaterialBorosilicate Glass
Label TypeMarking Spot
SterilityNon-sterile
GraduatedNo
Diameter (Metric)16 mm
Product TypeCulture Tube
Unit SizePK