Sterile Plastic Culture Tubes: Clear Polystyrene
Sterile Plastic Culture Tubes: Clear Polystyrene
Actual product may vary
Sterile Plastic Culture Tubes: Clear Polystyrene
Sterile Plastic Culture Tubes: Clear Polystyrene
Sterile Plastic Culture Tubes: Clear Polystyrene
Sterile Plastic Culture Tubes: Clear Polystyrene
Ideal for tissue culture procedures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Length (Metric)Closure TypePackaging
FIS14-956-5B16 mL125 mmScrew CapBlister Packs
FIS14-956-3C5 mL75 mmSnap CapBlister Packs
FIS14-956-3A5 mL75 mmNot IncludedBulk - 1 Pk. of 125
FIS14-956-5A125 mmScrew CapIndividually Wrapped, 500 Pks. of 1
FIS14-956-5C19 mL150 mmScrew CapIndividually Wrapped, 500 Pks. of 1
FIS14-956-6A14 mL100 mmSnap CapIndividually Wrapped, 500 Pks. of 1
FIS14-956-6C14 mL100 mmNot IncludedBulk - 8 Pks. of 125
Catalog number FIS14-956-5B
Price (SGD)
-
Capacity (Metric):
16 mL
Length (Metric):
125 mm
Closure Type:
Screw Cap
Packaging:
Blister Packs
Ideal for tissue culture procedures
  • Gamma-irradiated and tested for sterility; ready to use
  • No chance of residual contamination
  • All screw-cap tubes have outside threads and unconstricted necks, making filling and pipetting easier
  • All snap caps can be positioned for aerobic or anaerobic applications
Specifications
ColorClear
Capacity (Metric)16 mL
Length (Metric)125 mm
MaterialPolystyrene
For Use WithIdeal for tissue culture procedures
IncludesScrew cap
PackagingBlister Packs
SterilitySterile
Diameter (Metric)16 mm
Closure TypeScrew Cap
Product TypeCulture Tube
Unit SizeCS