Sterile Plastic Culture Tubes: Translucent Polypropylene
Sterile Plastic Culture Tubes: Translucent Polypropylene
Actual product may vary
Sterile Plastic Culture Tubes: Translucent Polypropylene
Sterile Plastic Culture Tubes: Translucent Polypropylene
Sterile Plastic Culture Tubes: Translucent Polypropylene
Sterile Plastic Culture Tubes: Translucent Polypropylene
Ideal for tissue culture procedures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Length (Metric)Closure TypePackaging
FIS14-956-1G14 mL100 mmNot IncludedBlister pack of 125, 1000/Cs.
FIS14-956-1C5 mL75 mmSnap CapIndividually wrapped, 500/Cs.
FIS14-956-1B5 mL75 mmNot IncludedBlister pack of 125, 1000/Cs.
FIS14-956-1D5 mL75 mmSnap CapBlister pack of 25, 500/Cs.
FIS14-956-1J14 mL100 mmSnap CapBlister pack of 25, 500/Cs.
Catalog number FIS14-956-1G
Price (SGD)
-
Capacity (Metric):
14 mL
Length (Metric):
100 mm
Closure Type:
Not Included
Packaging:
Blister pack of 125, 1000/Cs.
Ideal for tissue culture procedures
  • Gamma-irradiated and tested for sterility; ready to use
  • No chance of residual contamination
  • All snap caps can be positioned for aerobic or anaerobic applications
Specifications
ColorTranslucent
Capacity (Metric)14 mL
Length (Metric)100 mm
MaterialPolypropylene
For Use WithIdeal for tissue culture procedures
PackagingBlister pack of 125, 1000/Cs.
SterilitySterile
Diameter (Metric)17 mm
Closure TypeNot Included
Product TypeCulture Tube
Unit SizeCS