Sterile Sampling Bags with Flat-Wire Closures
Actual product may vary
Fisherbrand
Sterile Sampling Bags with Flat-Wire Closures
An economical and efficient way to collect, contain and carry samples
Have Questions?
Catalog NumberDimensions (L x W)Thickness (English)Capacity (English)Color
FIS14-955-1869 x 4.5 in. (22.9 x 11.4cm)3 mil18 oz.Clear
Catalog number FIS14-955-186
Price (SGD)
72.00
Pack
Dimensions (L x W):
9 x 4.5 in. (22.9 x 11.4cm)
Thickness (English):
3 mil
Capacity (English):
18 oz.
Color:
Clear
Price (SGD)
72.00
Pack
Collect, contain and carry samples in an economical and efficient way
  • Sterile to avoid sample contamination
Specifications
Closure TypeFlat Wire
ColorClear
MaterialPolyethylene
Thickness (English)3 mil
Dimensions (L x W)9 x 4.5 in. (22.9 x 11.4cm)
Length (English)9 in.
Width (English)4.5 in.
Capacity (English)18 oz.
Length (Metric)22.9 cm
Width (Metric)11.4 cm
Unit SizePack