PYREX™ Reusable Glass Tubes with Rubber-Lined Phenolic Caps
PYREX™ Reusable Glass Tubes with Rubber-Lined Phenolic Caps
Actual product may vary
PYREX™ Reusable Glass Tubes with Rubber-Lined Phenolic Caps
PYREX™ Reusable Glass Tubes with Rubber-Lined Phenolic Caps
PYREX™ Reusable Glass Tubes with Rubber-Lined Phenolic Caps
PYREX™ Reusable Glass Tubes with Rubber-Lined Phenolic Caps
Deep-form phenolic caps facilitate handling and sealing after autoclaving
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Length (Metric)Thread SizeCapacity (Metric)
FIS14-932D20 mm150 mm18-41534 mL
FIS14-932A16 mm125 mm15-41515 mL
FIS14-932B16 mm150 mm15-41520 mL
Catalog number FIS14-932D
Price (SGD)
1,114.00
PK
Add to cart
Diameter (Metric):
20 mm
Length (Metric):
150 mm
Thread Size:
18-415
Capacity (Metric):
34 mL
Price (SGD)
1,114.00
PK
Add to cart
Deep-form phenolic caps facilitate handling and sealing after autoclaving
Furnished with deep-form phenolic caps to facilitate handling and sealing after autoclaving. Caps are fabricated from a special formula resistant to the effects of temperature and steam in autoclaving. Caps are provided with a glued-in, white rubber liner. The white enamel marking spot gives an excellent surface for pencil notations. Pencil marks are easily erased.
For phenolic caps, see Cat. No. 9999.
Specifications
Closure ColorBlack
Closure MaterialPhenolic
Design TypeThreaded
Capacity (Metric)34 mL
Length (Metric)150 mm
MaterialBorosilicate Glass
Label TypeMarking Spot
Liner MaterialRubber
Thread Size18-415
SterilityNon-sterile
AutoclavableAutoclavable
GraduatedNo
Diameter (Metric)20 mm
Closure TypeScrew Cap
Product TypeCulture Tube
Unit SizePK