Kimble™ KIMAX™ Reusable Borosilicate Glass Tubes with Plain End and Marking Spot
Kimble™ KIMAX™ Reusable Borosilicate Glass Tubes with Plain End and Marking Spot
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Reusable Borosilicate Glass Tubes with Plain End and Marking Spot
With durable white ceramic marking spot
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Length (Metric)Capacity (Metric)
FIS14-923P25 mm150 mm55 mL
FIS14-923F15 mm125 mm16 mL
Catalog number FIS14-923P
Price (SGD)
471.00
PK
Add to cart
Diameter (Metric):
25 mm
Length (Metric):
150 mm
Capacity (Metric):
55 mL
Price (SGD)
471.00
PK
Add to cart
With durable white ceramic marking spot
  • Fire-polished top rim
  • Uniform wall thickness for maximum heat transfer and chemical resistance
  • Culture tubes made from KG-33 glass feature fire-polished top rim and uniform wall thickness for maximum heat transfer and chemical resistance
  • Designed from ASTM™ specification E982, Type IV requirements
Specifications
Certifications/ComplianceASTM E982, Type IV
Design TypePlain
Capacity (Metric)55 mL
Length (Metric)150 mm
MaterialBorosilicate Glass
Label TypeMarking Spot
Diameter (Metric)25 mm
Product TypeReusable Glass Tube
Unit SizePK