Kimble™ KIMAX™ Reusable Test Tubes with Beaded Rim
Kimble™ KIMAX™ Reusable Test Tubes with Beaded Rim
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Reusable Test Tubes with Beaded Rim
Made from N-51A borosilicate glass
Have Questions?
Catalog NumberDiameter (Metric)Length (Metric)Capacity (Metric)
FIS14-915G25 mm150 mm55 mL
Catalog number FIS14-915G
Price (SGD)
339.00
PK
Add to cart
Diameter (Metric):
25 mm
Length (Metric):
150 mm
Capacity (Metric):
55 mL
Price (SGD)
339.00
PK
Add to cart
  • Uniform wall thickness for maximum heat transfer and chemical resistance
Specifications
Certifications/ComplianceASTM E438, Type I, Class B
Design TypePlain Beaded Rim
Capacity (Metric)55 mL
Length (Metric)150 mm
MaterialBorosilicate Glass
Label TypeNone
Diameter (Metric)25 mm
Unit SizePK