Safe-T-Fill™ Capillary Blood Collection Systems: EDTA
Actual product may vary
RAM Scientific
Safe-T-Fill™ Capillary Blood Collection Systems: EDTA
No scooping needed, allowing for high-quality, tissue-free samples
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)AdditiveProduct Type
FIS14-915-86200 μLK2 EDTASelf-sealing Whole Blood Tube
FIS14-915-53300 μLK2 EDTAHematology/Whole Blood Tube
FIS14-915-85125 μLK2 EDTASelf-sealing Whole Blood Tube
Catalog number FIS14-915-86
Price (SGD)
-
Capacity (Metric):
200 μL
Additive:
K2 EDTA
Product Type:
Self-sealing Whole Blood Tube
Blood samples are drawn into the SAFE-T-FILL™ Plastic Micro Capillary Blood Collection Tubes through capillary action, reducing hemolysis,
  • 100% plastic tubes collect microliter volume capillary blood samples from finger and heel sticks
  • Blood samples are drawn into the microtube through capillary action, reducing hemolysis, tissue contamination, and re-draws. No "scooping" or "milking" required
  • Capillary straw and microtube are coated with anticoagulant Dipotassium (K2) EDTA (Purple cap) to prevent clotting
  • Attached cap is color-coded to designate the type of additive that is in both the capillary tube and the microtube
  • Lot Number, Expiration Date, and fill lines are printed directly on the microtube
  • A complete end-to-end fill of the capillary tube accurately measures the stated fill volume
  • For whole blood hematology determinations
  • Small sample sizes, great for pediatrics and neonates
  • Available with self-sealing rubber cap (Not made with natural rubber latex)
  • Available in a variety of sizes and draw volumes
Specifications
ColorClear
Product TypeSelf-sealing Whole Blood Tube
AdditiveK2 EDTA
MaterialPlastic
SterilityNon-sterile
Closure TypeSelf-sealing Pierceable Cap
Capacity (Metric)200 μL
Closure IncludedYes
Closure ColorPurple
Quantity50 Tubes/Bag, 10 Bags/Cs.
For Use With (Application)Intended for hematology (haematology) determinations that require whole blood samples. Self-sealing cap ensures automatic closure, facilitating transport and prevents contamination.
Unit SizePack