Check Order Status
Lead Acetate Test Paper
Actual product may vary
Lead Acetate Test Paper
Detects Hydrogen Sulfide in water in concentrations as low as 5ppm. Use this test paper to test your drinking water, well water or groundwater.
Have Questions?
Catalog Number
Quantity
FIS14-862
(24 vials, 100 strips/vial)
Catalog number
FIS14-862
Price (SGD)
152.00
PK
Add to cart
Quantity:
(24 vials, 100 strips/vial)
Price (SGD)
152.00
PK
Add to cart
Use this strip to test for the presence of hydrogen sulfide in concentrations as low as 5ppm
Specifications
For Use With (Application)
Use this strip to test for the presence of hydrogen sulfide
Includes
100 Test Strips/Vial
Quantity
(24 vials, 100 strips/vial)
Shelf Life
2 Years
Type
Lead Acetate Test Paper