Check Order Status
Arsenal™ 5960T Canvas Hoist Bucket with D-Rings and Top
Actual product may vary
Arsenal™ 5960T Canvas Hoist Bucket with D-Rings and Top
Arsenal™ 5960T Canvas Hoist Bucket with D-Rings and Top is constructed of a canvas main material and a synthetic leather bottom.
Have Questions?
Catalog number
FIS14-860
Price (SGD)
160.00
PK
Add to cart
Price (SGD)
160.00
PK
Add to cart
24 oz. canvas main material
Synthetic leather reinforced base
Molded plastic top ring to maintain wide opening for viewing and accessing contents
High tensile strength web handle with leather reinforcement
Zinc plated alloy steel d-ring on handle for hoisting
Zinc plated alloy steel d-rings built into handle ends for anchoring lanyards
Heavy-gauge nylon stitching
Interior pocket
Nickel-plated storm drain
Certified to a 5:1 safety factor
Specifications
Height (English)
17 in.
Height (Metric)
43 cm
Load Bearing Capacity (English)
150 lb.
Load Bearing Capacity (Metric)
68 kg
Size
Large
Color
Gray
Diameter (English)
12.5 in.
Diameter (Metric)
32 cm
Material
Canvas