Hydrion™ Sanitizer Test Kits: Quaternary Test Paper Kits
Actual product may vary
Micro Essential Laboratory
Hydrion™ Sanitizer Test Kits: Quaternary Test Paper Kits
Provides a simple, reliable and economical means to measure quaternary concentration of ammonium, chlorine and iodine sanitizer solutions
Have Questions?
Catalog NumberTypeRange
FIS14-853-180QT-40 Test for 4-chain Quats0 to 500 ppm
Catalog number FIS14-853-180
Price (SGD)
214.00
Case
Type:
QT-40 Test for 4-chain Quats
Range:
0 to 500 ppm
Price (SGD)
214.00
Case
Provides a simple, reliable and economical means to measure quaternary concentration of ammonium, chlorine and iodine sanitizer solutions
Federal, State and Local health regulations require users of Quaternary Ammonium Sanitizer Solutions to have appropriate test kits available to verify the strength of sanitizer solutions. Sanitizer solutions are essential in the food service industry to ensure that sanitizers are at the proper concentration specified by the individual manufacturer.
Hydrion Test for Two-chain Quats (FS Cat. No. 15-420-1A)
  • Measures 0 to 400ppm
  • Detects exhaustion of solutions that should be replaced, as well as helping to avoid using excessive amounts of sanitizing agents
  • Calibrated color chart
  • Color matches at 0-100-200-300-400ppm
  • Measures n-alkyldimethylbenzyl and/or n-alkyl dimethyl ethyl benzyl ammonium chloride and Roccall II
  • Each carton contains 10 kits, each consisting of a 15-foot roll of test paper and matching color chart
Specifications
For Use With (Application)To measure the concentration of quaternary sanitizers, particularly muli-quat broad range quaternary ammonium sanitizer solutions
IncludesOne 15 ft. roll of test paper, matching color chart
Quantity10 Case
Range0 to 500 ppm
TypeQT-40 Test for 4-chain Quats
Unit SizeCase