Hydrion™ One Drop Concentrated Indicator Solution Kit
Actual product may vary
Micro Essential Laboratory
Hydrion™ One Drop Concentrated Indicator Solution Kit
Concentrated pH indicator solution only needs one drop to test pH
Have Questions?
Catalog NumberpH Range
FIS14-853-1631 to 11
Catalog number FIS14-853-163
Price (SGD)
39.00
Each
pH Range:
1 to 11
Price (SGD)
39.00
Each
  • One drop of indicator solution added to 4mL (0.14 oz.) of sample provides accurate pH reading
  • Bright, easy to view color display remains stable for delayed readings
  • pH Range: 1.0 to 11.0
  • Color chart has distinct matches at every full pH unit
For testing pH of solutions
Specifications
pH Range1 to 11
For Use With (Application)For testing pH of solutions
Includes24mL of pH indicator solution in polyethylene dropper bottle , 4mL sample tube (capped) and a laminated color chart, packed in a polypropylene protective case
QuantityEach
TypepH Indicator Solution
Unit SizeEach