Single-Roll Hydrion™ pH Test Paper, With Chart
Actual product may vary
Single-Roll Hydrion™ pH Test Paper, With Chart
Single-Roll Hydrion™ pH Test Paper, With Chart
Micro Essential Laboratory
Single-Roll Hydrion™ pH Test Paper, With Chart
For measuring pH solutions.
Have Questions?
Catalog NumberpH Range
FIS14-853-150R4 to 9
Catalog number FIS14-853-150R
Price (SGD)
18.00
Each
pH Range:
4 to 9
Price (SGD)
18.00
Each
  • Used in laboratories, hospitals and universities worldwide
  • Comes with a 7/32 in. x 15 ft. roll of test paper and matching color chart
  • Provides for approximately 100 tests
Specifications
pH Range4 to 9
For Use With (Application)For classroom demonstrations, hospital laboratories, general laboratories, industrial testing, food service, safety testing, emergency testing, agricultural testing
IncludesColorimetric chart
Quantity100 Tests
TypepH Test Paper
Unit SizeEach