Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
pH and Electrochemistry
▸
pH Paper and other Test Strips
▸
Insta-Chek™ 0-13 Wide-Range pH Test Paper/Dispenser
Actual product may vary
Micro Essential Laboratory
Insta-Chek™ 0-13 Wide-Range pH Test Paper/Dispenser
Direct reading pH paper with 14 colors for 14 pH values—the widest range available on a single paper
Have Questions?
Catalog Number
Format
Length (English)
Quantity
FIS14-850-2
Jumbo Roll
15 ft.
1000 Tests
Catalog number
FIS14-850-2
Price (SGD)
35.00
Each
Add to cart
Format:
Jumbo Roll
Length (English):
15 ft.
Quantity:
1000 Tests
Price (SGD)
35.00
Each
Add to cart
Direct reading pH paper with 14 colors for 14 pH values—the widest range available on a single paper
Immediate response—colors are distinct and easily read for accurate, reproducible readings
Single roll dispenser with one 50 ft. × 0.5 in. roll
Specifications
pH Range
0 to 13
Format
Jumbo Roll
Includes
Colorimetric chart
Quantity
1000 Tests
Length (English)
15 ft.
Length (Metric)
4.57 m