Acid/Alkali Test Papers
Actual product may vary
Acid/Alkali Test Papers
Acid/Alkali Test Papers
Acid/Alkali Test Papers
Acid/Alkali Test Papers
Acid/Alkali Test Papers
Acid/Alkali Test Papers
Acid/Alkali Test Papers
Acid/Alkali Test Papers
Acid/Alkali Test Papers
Acid/Alkali Test Papers
Foolproof pH test to determine if a solution is acidic or alkaline.
Have Questions?
Catalog NumberpH RangeType
FIS14-8500 to 14Litmus Neutral Test Strip
Catalog number FIS14-850
Price (SGD)
81.00
Pack
pH Range:
0 to 14
Type:
Litmus Neutral Test Strip
Price (SGD)
81.00
Pack
  • A foolproof pH test to determine if a solution is acidic
  • The Blue Litmus test paper does not come with a color chart as it cannot provide you with a defined pH value
  • The test strip will turn red if dipped into an acidic solution (pH <7)
  • It will remain unchanged in an alkaline solution
  • We have tested the blue litmus with multiple pH buffers
  • The blue paper is blue at pH 8
  • At pH 7, the sample is maybe just a little red
  • The real change to red seems to start with pH 6
  • 5 and lower
  • Neutral Litmus test paper tests for Neutrality of a solution, or its acidity or alkalinity
  • The test strip will turn red if dipped into an acidic solution (pH <7) or blue if dipped into an alkaline solution (pH >7)
  • Red Litmus test paper is a foolproof pH test to determine if a solution is alkaline
  • The test strip will turn blue if dipped into an alkaline solution (pH >7)
  • Color chart not included
Specifications
Certifications/ComplianceOSHA, 29 CFR 1910.133, EN166
pH Range0 to 14
Quantity24 x 100 Strips
TypeLitmus Neutral Test Strip
Unit SizePack