Legato 100 Infuse Only Syringe Pump
Actual product may vary
KD Scientific
Legato 100 Infuse Only Syringe Pump
Versatile pump offers unparalleled ease of use through simple touch-screen interface
Have Questions?
Catalog NumberFlow Rate
FIS14-831-2101.28 pL/min. to 88.28 mL/min.
Catalog number FIS14-831-210
Price (SGD)
7,450.00
Each
Flow Rate:
1.28 pL/min. to 88.28 mL/min.
Price (SGD)
7,450.00
Each
Versatile pump offers unparalleled ease of use through simple touch-screen interface
Infuse only single syringe pump. Accommodates syringes from 0.5μL to 60mL. User definable flowrates with selectable target volume or time values to control the total infusion volume.
  • Holds one syringe from 0.5μL to 60mL
  • High resolution color touch screen
  • Touch pad lock feature
  • LED light on front panel
  • Full metal chassis
  • Built-in syringe table
  • Up to 30 lb. linear force
  • Advanced microstepping techniques
  • Built-in RS485 interface
  • USB port
  • I/O and TTL interface
  • Continuous mode of operation
  • Spill dam
Specifications
  • Accuracy: ±0.5 %
  • Reproducibility: ±0.05%
  • Syringes (min/max): 0.5μL to 60mL
  • Flow Rate, Minimum (0.5μL syringe): 1.28 pL/min.
  • Flow Rate, Maximum (10mL syringe): 25.99 mL/min.
  • Flow Rate, Maximum (60mL syringe): 88.28 mL/min.
  • Display: 4.3 WQVGA TFT Color Display with touchpad
  • Non-Volatile Memory: Stores all settings
  • Operating Temperature: 4°C to 40°C (40°F to 104°F)
  • Storage Temperature: -10°C to 70°C (14°F to 158°F)
  • Humidity: 20% to 80% RH noncondensing
  • Mode of Operation: Continuous
  • Power: 100-240 VAC: 50/60 Hz, 50W, 0.5A fuse
  • Dimensions: 9 x 7.5 x 5 in. (22.6 x 19.05 x 15cm)
  • Weight: 5.9 lb. (2.66kg)
Specifications
Voltage100/240 V
Flow Rate1.28 pL/min. to 88.28 mL/min.
Capacity (Metric)0.5 μL to 60 mL
Certifications/ComplianceCE, UL, CSA, CB Scheme, EU RoHS
ConnectorsRS485 - IEEE-1394 6 pos, USB - Type B, I/O and TTL - 15 pin D-sub connector
ControllerMicroprocessor with 0.062 microstepping
Linear Force (Metric)13.6 kg
Linear Force (English)30 lb.
MaterialMetal
Reproducibility±0.05%
Weight (English)5.9 lb.
Weight (Metric)2.66 kg
Electrical Requirements100/240 V 50/60 Hz
Display TypeTFT
Flow Rate Accuracy±0.5%
Number of Channels1
Frequency50/60 Hz
Overall Height (English)5 in.
Overall Height (Metric)15 cm
Overall Length (English)9 in.
Overall Length (Metric)7.5 cm
Overall Width (English)7.5 in.
Temperature (Metric) Max40°C
Temperature (Metric) Min.4°C
Product TypeInfusion Syringe Pump
Storage Requirements-10°C to +70°C
Unit SizeEach