▸
First Aid and Medical
▸
Sharps Disposal Containers and Accessories
▸
In-Room™ Sharps Disposal System with Mailbox-Style Lids
Actual product may vary
Covidien
In-Room™ Sharps Disposal System with Mailbox-Style Lids
Provides an economical horizontal drop with a closable lid
Have Questions?
Catalog Number
Capacity (English)
FIS14-827-55B
2 qt.
Catalog number
FIS14-827-55B
Price (SGD)
-
Capacity (English):
2 qt.
Mailbox-style lid can be closed when container is not in use
Economical horizontal drop maximizes use of container volume
Containers lock to make them secure for final disposal
Specifications
Capacity (Metric)
1.9 L
Closure Type
Mailbox-Style Lid, Lockable
Capacity (English)
2 qt.
Color
Translucent Red
Quantity
20/Cs.
Autoclavable
Autoclavable
Material
Polypropylene