700 Microliter Syringes: N Termination
Actual product may vary
Hamilton
700 Microliter Syringes: N Termination
The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 500 μL. A variety of different terminations and needle options are available in this series.
Have Questions?
Catalog NumberVolume (Metric)Needle Point StyleDiameter (Metric) Inner NeedleDiameter (Metric) Outer Needle
FIS14-824-6100 μLSharp, Bevel, Curved, Non-coring0.15 mm0.72 mm
Catalog number FIS14-824-6
Price (SGD)
-
Volume (Metric):
100 μL
Needle Point Style:
Sharp, Bevel, Curved, Non-coring
Diameter (Metric) Inner Needle:
0.15 mm
Diameter (Metric) Outer Needle:
0.72 mm
  • Original hand-fitted Hamilton syringe
  • Ideal for dispensing volumes from 0.5 µL up to 500 µL
  • A variety of different terminations and needle options are available in this series
Specifications
AutoclavableNot Autoclavable
MaterialStainless Steel (Plunger)
Needle Gauge22s G
Diameter (English) Inner0.061 in.
Diameter (English) Inner Needle0.0059 in.
Diameter (Metric) Inner Needle0.15 mm
Diameter (English) Outer Needle0.0283 in.
Diameter (English) Outer0.305 in.
Diameter (Metric) Inner1.46 mm
Diameter (Metric) Outer7.75 mm
Diameter (Metric) Outer Needle0.72 mm
For Use With (Application)For dispensing low volumes to achieve the highest level of accuracy and precision possible
Length (English)2 in.
Length (English) Needle2 in.
Length (Metric)51 mm
Product TypeMicroliter Syringe
Max. Pressure2000 psi
Max. Temperature (English)122°F
Max. Temperature (Metric)50°C
Min. Temperature (English)50°F
Model710
Needle Point Number2
QuantityEa.
Volume (Metric)100 μL
Needle Point StyleSharp, Bevel, Curved, Non-coring
Connection TypeCemented
Unit SizeEach