▸
Chromatography Syringes
▸
Chromatography Syringe Accessories
▸
Tungsten Cleaning Wires
Actual product may vary
Hamilton
Tungsten Cleaning Wires
For mechanical cleaning to remove needle obstructions.
Have Questions?
Catalog Number
For Use With (Equipment)
FIS14-819-76
22s, 25s and 28 to 30G Needle, Syringe and Needle Cleaning Kit
Catalog number
FIS14-819-76
Price (SGD)
-
For Use With (Equipment):
22s, 25s and 28 to 30G Needle, Syringe and Needle Cleaning Kit
Can be threaded through the needle to clear blockages
For a variety of needle gauges
Provided in packs of 10
Specifications
Material
Tungsten
Diameter (Metric) Outer
0.126 mm
Length (English)
7 in.
Length (Metric)
17.78 cm
Product Type
Cleaning Wire
Packaging
10/Pk.
For Use With (Equipment)
22s, 25s and 28 to 30G Needle, Syringe and Needle Cleaning Kit