▸
First Aid and Medical
▸
Patient Care Products
▸
Medical Syringes
▸
HSW Soft-Ject™ Disposable Syringes
Actual product may vary
Air-Tite
HSW Soft-Ject™ Disposable Syringes
High-quality, three-part disposable rubber piston syringes
Have Questions?
Change view
Catalog Number
Volume (Metric)
FIS14-817-54
10 mL
FIS14-817-57
50 mL
FIS14-817-53
5 mL
3 Options
Catalog number
FIS14-817-54
Price (SGD)
-
Volume (Metric):
10 mL
High-quality, three-part disposable rubber piston syringes
Lubricated synthetic rubber piston allows for smooth draw while positive stop helps prevent accidental pullout
Bold graduations are easily read and resist wipe off
ETO gas sterilized syringes, latex and pyrogen free
Packaged in individual sterile strip packs
Made in Germany
Specifications
Certifications/Compliance
FDA, CE, ISO 9001
Latex Free
Yes
Product Type
Soft-Ject™ Syringe
Disposable
Single-use
Graduations
12 mL
Volume (Metric)
10 mL
Connection Type
Luer Lock
Sterility
Sterile