Premium Hypodermic Needles for Lab/Vet Use
Actual product may vary
Air-Tite
Premium Hypodermic Needles for Lab/Vet Use
Sterile, disposable needles for laboratory and veterinary use
Have Questions?
Catalog NumberNeedle GaugeLength (English) Needle
FIS14-817-10222 G4 in.
Catalog number FIS14-817-102
Price (SGD)
200.00
Pack
Needle Gauge:
22 G
Length (English) Needle:
4 in.
Price (SGD)
200.00
Pack
Ideal for laboratory and veterinary use; sterile and disposable.
  • Stainless-steel
  • Regular-wall design needles; all have 12° regular bevel and tri-bevel points
  • High-density polypropylene hub fits any Luer-Lok™ or Luer-Slip syringe tip
  • Come in strips of individual blister packs
Specifications
MaterialHDPP (Hub), Stainless Steel
Needle Gauge22 G
Diameter (English) Outer0.0285 in.
Diameter (Metric) Outer0.72 mm
Length (English) Needle4 in.
Product TypeHypodermic Needle
DisposableSingle-use
Length (Metric) Needle101.6 mm
Needle Point Style12° Regular Bevel and Tri-bevel Points
Connection TypeLuer Lock, Luer Slip