AutoclavableAutoclavable
MaterialNickel-plated Brass (Hub), Stainless Steel
Needle Gauge18 G
Diameter (English) Inner0.033 in.
Diameter (English) Outer0.05 in.
Diameter (Metric) Inner0.84 mm
Diameter (Metric) Outer1.27 mm
Length (English) Needle2 in.
Product TypeNeedle
Needle Point StyleBlunt
For Use With (Equipment)PTFE Luer-lock Syringes and Luer-tip or PTFE Luer-lock Connectors
Unit SizePack