Nalgene™ Polypropylene-Filled Test Tube Peg Racks
Actual product may vary
Nalgene™ Polypropylene-Filled Test Tube Peg Racks
Nalgene™ Polypropylene-Filled Test Tube Peg Racks
Nalgene™ Polypropylene-Filled Test Tube Peg Racks
Thermo Scientific
Nalgene™ Polypropylene-Filled Test Tube Peg Racks
Hold sample collection tubes, reaction tubes and test tubes with these polypropylene test tube peg racks. Suitable for drying glass plates and chromatography plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorFor Use With (Equipment)
FIS14-810-54GWhite14-17mm Tubes
5977-0013White10-13mm Tubes
FIS14-810-54AWhite10-13mm Tubes
FKS10248680White14-17mm Tubes
5977-0313Blue10-13mm Tubes
FIS14-810-54DBlue10-13mm Tubes
FKS10763481Blue10-13mm Tubes
FIS14-810-54KBlue14-17mm Tubes
Catalog number FIS14-810-54G
Price (SGD)
126.00
Pack
Color:
White
For Use With (Equipment):
14-17mm Tubes
Price (SGD)
126.00
Pack
Hold sample collection tubes, reaction tubes and test tubes with Thermo Scientific™ Nalgene™ Filled Polypropylene Test Tube Peg Racks. Racks are suitable for drying glass plates used in electrophoresis, as well as chromatography plates. These chemically resistant and autoclavable racks will not float in a water bath.
  • Consist of an array of pegs spaced to hold given-diameter test tubes while allowing full view of the tube contents
  • Pegs also hold tubes in inverted position for drying
  • When inverted, ribs keep tubes off base to allow drainage through holes and minimize collection of airborne contaminants inside the tubes
  • Suitable for drying glass plates used in electrophoresis, as well as chromatography plates
  • Higher specific gravity than water so stays submerged in a water bath — won't float
  • Autoclavable
Please recycle the corrugated cartons and LDPE bags in which your Nalgene products are packaged.
Specifications
DescriptionTest Tube Rack Peg Wht PP 17mm
AutoclavableAutoclavable
ColorWhite
HoldsTubes (14 to 17 mm)
MaterialFilled Polypropylene
Positions50
ShapeRectangular
For Use With (Equipment)14-17mm Tubes
No. per Case8
No. per Pack2
Dimensions (L x W x H)7.3 x 4.1 x 2.75 in. (18.7 x 10.5 x 7 cm)
Length (Metric)18.7 cm
Length (English)7.3 in.
Width (English)4.1 in.
Height (English)2.75 in.
Diameter (Metric) Opening17 mm
Product TypeTest Tube Peg Rack
Diameter (Metric) Inner17 mm
Height (Metric)7 cm
Array5 x 10
Width (Metric)10.5 cm
Unit SizePack