Three-Tier Polypropylene Test Tube Racks
Three-Tier Polypropylene Test Tube Racks
Actual product may vary
Three-Tier Polypropylene Test Tube Racks
Molded racks provide rigid support when fully loaded—reduce breakage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHolds
FIS14-809DTubes (25 to 30 mm)
FIS14-809ATubes (10 to 13 mm)
FIS14-809AATubes (13 to 16 mm)
FIS14-809BTubes (16 to 20 mm)
FIS14-809CTubes (20 to 25 mm)
Catalog number FIS14-809D
Price (SGD)
125.00
EA
Add to cart
Holds:
Tubes (25 to 30 mm)
Price (SGD)
125.00
EA
Add to cart
Molded racks provide rigid support when fully loaded—reduce breakage.
  • Stack empty or loaded with tubes up to 10 cm (4 in.) long
  • Autoclavable to 121°C (250°F)
  • With convenient end plate handles for carrying
  • Rows are numbered and lettered for easy referencing
  • White surfaces make color comparison easy
  • Sturdy and chemical-resistant, will not shatter or stain
Specifications
Product TypeTube Rack
Array3 X 8
AutoclavableAutoclavable
ColorWhite
HoldsTubes (25 to 30 mm)
MaterialPolypropylene
Positions24
ShapeRectangular
Shelves3 Tiers
StackableYes
SterilityNon-sterile
Dimensions (L x W x H)11.69 x 5 x 5.25 in. (29.7 x 12.7 x 13.3 cm)
Length (Metric)29.7 cm
Length (English)11.69 in.
Width (Metric)12.7 cm
Width (English)5 in.
Height (Metric)13.3 cm
Height (English)5.25 in.
Diameter (Metric) Opening30 mm
Diameter (Metric) Inner30 mm
Unit SizeEA