Three-Tier Polypropylene Test Tube Racks
Actual product may vary
Three-Tier Polypropylene Test Tube Racks
Molded racks provide rigid support when fully loaded—reduce breakage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHolds
FIS14-809BTubes (16 to 20 mm)
FIS14-809DTubes (25 to 30 mm)
FIS14-809ATubes (10 to 13 mm)
FIS14-809AATubes (13 to 16 mm)
FIS14-809CTubes (20 to 25 mm)
Catalog number FIS14-809B
Price (SGD)
83.00
Each
Holds:
Tubes (16 to 20 mm)
Price (SGD)
83.00
Each
Molded racks provide rigid support when fully loaded—reduce breakage.
  • Stack empty or loaded with tubes up to 10 cm (4 in.) long
  • Autoclavable to 121°C (250°F)
  • With convenient end plate handles for carrying
  • Rows are numbered and lettered for easy referencing
  • White surfaces make color comparison easy
  • Sturdy and chemical-resistant, will not shatter or stain
Specifications
AutoclavableAutoclavable
ColorWhite
HoldsTubes (16 to 20 mm)
MaterialPolypropylene
Positions40
ShapeRectangular
StackableYes, When Empty
SterilityNon-sterile
Dimensions (L x W x H)10.63 x 4.44 x 5.25 in. (27 x 11.3 x 13.3 cm)
Length (Metric)27 cm
Length (English)10.63 in.
Width (English)4.44 in.
Height (English)5.25 in.
Diameter (Metric) Opening20 mm
Product TypeTube Rack
Diameter (Metric) Inner20 mm
Shelves3 Tiers
Height (Metric)13.3 cm
Array4 x 10
Width (Metric)11.3 cm
Unit SizeEach