Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology
Actual product may vary
Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology
Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology
Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology
Thermo Scientific
Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology
Securely hold a full load of test tubes or centrifuge tubes for 13, 16, 20, 25 and 30mm diameter sizes with these strong, durable racks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorArrayFor Use With (Equipment)
FIS14-809-38Yellow4 x 1020mm Tubes
5970-0513Red6 x 1213mm Tubes
FIS14-809-60Red6 x 1213mm Tubes
5970-0213Yellow6 x 1213mm Tubes
5970-0113Orange6 x 1213mm Tubes
5970-0013White6 x 1213mm Tubes
FIS14-809-22White6 x 1213mm Tubes
FIS14-809-62Red6 x 1216mm Tubes
FKS10435071Red6 x 1216mm Tubes
5970-0316Blue6 x 1216mm Tubes
FIS14-809-43Blue6 x 1216mm Tubes
FKS10098210Blue6 x 1216mm Tubes
5970-0416Green6 x 1216mm Tubes
FKS10343461Yellow6 x 1216mm Tubes
5970-0116Orange6 x 1216mm Tubes
FIS14-809-24White6 x 1216mm Tubes
5970-0520Red4 x 1020mm Tubes
FIS14-809-64Red4 x 1020mm Tubes
5970-0320Blue4 x 1020mm Tubes
FIS14-809-52Blue4 x 1020mm Tubes
5970-0020White4 x 1020mm Tubes
FIS14-809-26White4 x 1020mm Tubes
FKS10291761White4 x 1020mm Tubes
FIS14-809-66Red4 x 1025mm Tubes
FKS10663721Red4 x 1025mm Tubes
5970-0425Green4 x 1025mm Tubes
5970-0530Red3 x 830mm Tubes
FIS14-809-68Red3 x 830mm Tubes
5970-0330Blue3 x 830mm Tubes
FIS14-809-44Blue3 x 830mm Tubes
FKS10778721Blue3 x 830mm Tubes
5970-0430Green3 x 830mm Tubes
FIS14-809-49Green3 x 830mm Tubes
5970-0230Yellow3 x 830mm Tubes
FIS14-809-50Yellow3 x 830mm Tubes
FIS14-809-36Orange3 x 830mm Tubes
5970-0030White3 x 830mm Tubes
FIS14-809-30White3 x 830mm Tubes
FKS10644101White3 x 830mm Tubes
Catalog number FIS14-809-38
Price (SGD)
103.00
Each
Color:
Yellow
Array:
4 x 10
For Use With (Equipment):
20mm Tubes
Price (SGD)
103.00
Each
Securely hold a full load of test tubes or centrifuge tubes for 13, 16, 20, 25 and 30mm diameter sizes with Thermo Scientific™ Nalgene™ Unwire™ Test Tube Racks. Molded in a single piece for exceptional strength and durability, these racks have no coating to chip, peel or rust.
  • Smooth finish leaves fewer places for contaminants to lodge; no welds to break, no sharp edges.
  • Endplates accept large labels for easy identification of samples.
  • Higher specific gravity than water so it stays submerged in a water bath and won't float.
  • Withstands most lab chemicals and temperatures up to 121°C for a variety of lab procedures.
  • Available in six bright colors to help keep samples organized.
  • 15mL conical tubes fit 20mm racks; 50mL conical fit 30mm racks.
  • No assembly is required; rack is ready to use.
Please recycle the corrugated cartons and LDPE bags in which your Nalgene products are packaged.
Specifications
DescriptionTest Tube Rack Unwire™ Yellow 20mm
AutoclavableAutoclavable
ColorYellow
HoldsTubes (20 mm)
MaterialRESMER
Positions40
ShapeRectangular
For Use With (Equipment)20mm Tubes
Dimensions (L x W x H)9.75 x 4 x 3.25 in. (25 x 10.2 x 8.3 cm)
Length (Metric)25 cm
Length (English)9.75 in.
Width (English)4 in.
Height (English)3.25 in.
Diameter (Metric) Opening20 mm
Product TypeTest Tube Rack
Diameter (Metric) Inner20 mm
Shelves3 Tiers
Height (Metric)8.3 cm
Array4 x 10
Width (Metric)10.2 cm
Unit SizeEach