Two-Tier Polypropylene Test Tube Racks
Actual product may vary
Two-Tier Polypropylene Test Tube Racks
Fit most water baths for elevated temperature incubations.
Have Questions?
Catalog NumberPositionsHolds
FIS14-809-2140Tubes (16 to 20 mm)
Catalog number FIS14-809-21
Price (SGD)
113.00
Each
Positions:
40
Holds:
Tubes (16 to 20 mm)
Price (SGD)
113.00
Each
Fit most water baths for elevated temperature incubations.
  • Just the right size for serum vials (10-13 mm diameter), RIA vials and T3/T4 reaction vials (13-16 and 16-20 mm diameter), scintillation vials (25-30 mm diameter), or any other vial or tube of same dimensions
  • Long-lasting and corrosion-resistant
  • End plates handles
  • Polypropylene; steam autoclavable at 121°C (250°F)
Specifications
AutoclavableAutoclavable
ColorWhite
HoldsTubes (16 to 20 mm)
MaterialPolypropylene
Positions40
ShapeRectangular
StackableNo
SterilityNon-sterile
Dimensions (L x W x H)10 x 4.37 x 3.5 in. (25.4 x 11 x 8.9 cm)
Length (Metric)25.4 cm
Length (English)10 in.
Width (English)4.37 in.
Height (English)3.5 in.
Diameter (Metric) Opening20 mm
Product TypeTube Rack
Diameter (Metric) Inner20 mm
Shelves2 Tiers
Height (Metric)8.9 cm
Array10 x 4
Width (Metric)11 cm
Unit SizeEach