Four-Funnel Support Arm
Four-Funnel Support Arm
Actual product may vary
Four-Funnel Support Arm
Clamps to rods and lattices
Have Questions?
Catalog NumberPositions
FIS14-740Q4
Catalog number FIS14-740Q
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Positions:
4
  • Wooden support
  • With metal clamp and setscrew for firm mounting
  • Support holes are slotted for easy manipulation of funnels
  • End holes have 1.12 in. (2.8cm) diameter
  • Center holes have 0.62 in. (1.6cm) diameter
  • All holes tapered at 60° angle
Specifications
Product TypeFunnel Support Arm
AutoclavableNot Autoclavable
HoldsFunnels
MaterialWood
Positions4
For Use With (Equipment)Funnel
Dimensions (L x W x H)13.75 x 2.37 x 0.56 in. (35 x 6 x 1.4cm)
Length (Metric)35 cm
Length (English)13.75 in.
Width (Metric)6 cm
Width (English)2.37 in.
Height (Metric)1.4 cm
Height (English)0.56 in.
Unit SizeEA