Heavy-Duty Rectangular Cast-Iron Support
Heavy-Duty Rectangular Cast-Iron Support
Actual product may vary
Heavy-Duty Rectangular Cast-Iron Support
For maximum stability and molded polyethylene tray for spillage protection
Have Questions?
Catalog number FIS14-679Q
Price (SGD)
0.00
EA
Add to cart
Price (SGD)
0.00
EA
Add to cart
For maximum stability and molded polyethylene tray for spillage protection
  • Concentric rings on tray provide convenient guide for positioning containers
  • Height to top of rod is 35 in. (89cm)
  • With steel rod and rubber feet to prevent sliding
Specifications
MaterialCast Iron
Length (English) Base9.75 in.
Length (Metric) Base25 cm
Width (English) Base9.18 in.
Width (Metric) Base23 cm
IncludesSteel rod and rubber feet to prevent sliding
Product TypeHeavy Duty Support
ShapeRectangular
Quantity1 Ea.
Unit SizeEA