▸
Clamps and Supports
▸
Lattice Supports
▸
Flexaframe™ Castaloy™ Hook Connector
Actual product may vary
Flexaframe™ Castaloy™ Hook Connector
Locks two Flexframe rods at a 90° angle
Have Questions?
Catalog Number
Product Type
FIS14-666-18Q
Hook Connector
Catalog number
FIS14-666-18Q
Price (SGD)
-
Product Type:
Hook Connector
Can't slip
Install without disassembling frame
Supplied with Flexframe sets
Holders work with 13mm diameter rods
Specifications
Product Type
Hook Connector
For Use With (Application)
Locks two rods at 90° angle, slip proof, install without disassembling frame, comes with flexaframe sets