Tight-Ties™
Tight-Ties™
Actual product may vary
Tight-Ties™
Works with all types of plastic and rubber tubing
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
FIS14-649122Cable Tie
FIS14-649-122Cable Tie
Catalog number FIS14-649122
Price (SGD)
40.00
PK
Add to cart
Type:
Cable Tie
Price (SGD)
40.00
PK
Add to cart
Works with all types of plastic and rubber tubing
  • Gas-sterilized
  • Self-locking
  • Chemical-corrosion-resistant
  • Rustproof
  • Non-conducting nylon
  • Unique tapered tips for fast inserting and tightening
Specifications
  • Usable within temperature range -40° to +185°F (-40° to +85°C)
  • Tensile strength: 12 to 50 lb. (5.4 to 22.7kg)
  • When fastened, the diameters adjust 2.60 to 101.60mm).(5/32 to 4 in.)
14-649122 Cable Tie 279.4mm (11 in.)
06-662-58 Cable Tie Assortment Pack Variety of lengths from 101.6 to 355.6mm (4 to 14 in.)
Specifications
TypeCable Tie
MaterialNylon
Description11 in. (279.4mm)
Length (Metric)279.4 mm
Unit SizePK