MOLYKOTE™ High-Vacuum Grease
Actual product may vary
Corning
MOLYKOTE™ High-Vacuum Grease
Designed for sealing and lubricating vacuum and pressure systems.
Have Questions?
Catalog NumberVolume (Metric)Volume (English)
FIS14-635-5D100 g3.53 oz.
Catalog number FIS14-635-5D
Price (SGD)
91.00
Each
Volume (Metric):
100 g
Volume (English):
3.53 oz.
Price (SGD)
91.00
Each
Designed for sealing and lubricating vacuum and pressure systems
  • Low vapor pressure, stiff, nonmelting silicone lubricant maintains its consistency from -40° to +204°C (-40° to +400°F)
  • Resists oxidation and chemicals; noncarbonaceous
Specifications
Product TypeHigh Vacuum Grease
For Use With (Application)Designed for sealing and lubricating vacuum and pressure system
For Use With (Equipment)Vacuum and Pressure Systems
Certifications/ComplianceFDA 21 CFR 175.300, USDA, NSF 61
ColorWhite
MaterialSilicone
PackagingTube
Temperature (English)-40°F to 400°F
Temperature (Metric)-40°C to +204°C
Volume (English)3.53 oz.
Volume (Metric)100 g
Unit SizeEach