Magnetic Stirring Bars
Magnetic Stirring Bars
Actual product may vary
Magnetic Stirring Bars
Magnetic Stirring Bars
Magnetic Stirring Bars
Fisherbrand
Magnetic Stirring Bars
Stir bar assortments come in a convenient storage box. Fisherbrand™ Magnetic Stirring Bars contain PTFE encapsulated Alnico magnets and come in various sizes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityShapeColor
FIS14-512-12912OctagonWhite
FIS14-512-13022SphereWhite
FIS14-512-13118PolygonWhite
FIS14-512-13224OctagonYellow, Red, Blue
FIS14-512-13312EggYellow, Red, Blue
Catalog number FIS14-512-129
Price (SGD)
105.00
Each
Add to cart
Quantity:
12
Shape:
Octagon
Color:
White
Price (SGD)
105.00
Each
Add to cart
Stir bar assortments come in a convenient storage box. Fisherbrand™ Magnetic Stirring Bars contain PTFE encapsulated Alnico magnets and come in various sizes.
  • Choose from Octagon, Cylindrical, Polygon, Micro, Egg-Shaped or Spinplus™ shaped assortments
  • FDA Grade, USP Class VI PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclavable
Specifications
AutoclavableAutoclavable
Certifications/ComplianceISO 9001:2008
ColorWhite
IncludesTwo each of the following sizes 0.5 x 0.31 in., 0.63 x 0.31 in., 1 x 0.38 in., 1.5 x 0.38 in., 2 x 0.38 in., and 2.5 x 0.38 in.
CoatingPTFE
Diameter (Metric)7.9 mm, 9.7 mm
Length (Metric)12.7 mm, 16 mm, 25.4 mm, 38 mm, 51 mm, 63.5 mm
MaterialAlnico Magnet
Quantity12
ShapeOctagon
TypeStir Bar
Unit SizeEach