Pitched Blade Propeller, Blade Only
Actual product may vary
Caframo
Pitched Blade Propeller, Blade Only
Stainless steel Pitched Blade Propeller for use with Caframo™ and Fisherbrand™ Overhead Stirrers and shafts.
Have Questions?
Catalog NumberDiameter (Metric)Includes
FIS14-501-07950 mm8 mm Bore, Set Screw
Catalog number FIS14-501-079
Price (SGD)
236.00
Each
Diameter (Metric):
50 mm
Includes:
8 mm Bore, Set Screw
Price (SGD)
236.00
Each
  • Electro-polished stainless steel 316L
  • Chemically inert and corrosion resistant 316L stainless steel
  • For low shear, high top-to-bottom pumping capacity
  • Scale up advantage if propeller used in production
Specifications
Diameter (English)2 in.
Diameter (Metric)50 mm
For Use With (Equipment)Caframo Overhead Stirrer
Includes8 mm Bore, Set Screw
ModelA531
TypePropeller Blade
Weight (Metric)12.9 g
MaterialElectropolished Stainless Steel (316)
Weight (English)0.46 oz.
Unit SizeEach