RealTorqueDigital™ Overhead Stirrer
Actual product may vary
RealTorqueDigital™ Overhead Stirrer
RealTorqueDigital™ Overhead Stirrer
Thermo Scientific
RealTorqueDigital™ Overhead Stirrer
RealTorqueDigital™ Overhead Stirrer is a high-torque, digital overhead stirrer offering speed and torque capabilities for a wide range of volumes and viscosities.
Have Questions?
Catalog NumberSpeed
FIS14-500-21020 to 3000 rpm
Catalog number FIS14-500-210
Price (SGD)
5,495.00
Each
Speed:
20 to 3000 rpm
Price (SGD)
5,495.00
Each
  • Stirrer handles high-volume/high-viscosity stirring as easily as homogenizing and dispersing - especially useful for continuous use
  • User-friendly keypad permits precise speed adjustment and digital LCD indicates rpm or torque to within ±1%
  • Electronic controls permit speed adjustment to single-digit increments and maintain set speed within 1% as solution viscosity changes
  • Rugged, sealed metal housing encloses the high-performance brushless DC motor with automatic overload protection; and two-speed transmission that electronically shifts between high-torque and high-speed range-actually transferring motor's power to chuck with 95% efficiency
  • Chuck is adjustable up to 10 mm (3/8 in.)
  • Chuck guard protects chuck from splashes and prevents accidental movement of chuck
  • Stirrer features electronic controls
Specifications
Speed20 to 3000 rpm
Capacity60 L
DisplayDigital
Voltage120 V
Wattage150 w
Dimensions (L x W x H)8 x 3.7 x 9 in. (20.3 x 9.5 x 22.9 cm)
Height (English)9 in.
Height (Metric)22.9 cm
Length (English)8 in.
Length (Metric)20.3 cm
Width (English)3.7 in.
Width (Metric)9.5 cm
Electrical Requirements120 V, 50/60 Hz
Frequency50/60 Hz
Horsepower1/5 hp
Certifications/ComplianceCSA
Chuck Size13 mm
IncludesChuck, chuck key, chuck guard, support rod and power cord
MaterialCast Aluminum
ModelDigital overhead stirrer
Motor TypeBrushless DC
No. of Speed Gears2
Torque30 in-lb.
TypeOverhead Stirrer
Viscosity50,000 mPas
Weight (English)11 lb.
Weight (Metric)5 kg
Unit SizeEach