▸
Shakers
▸
Wrist Action Shakers
▸
Wrist Action™ Model 75 Laboratory Shakers
Actual product may vary
Burrell Scientific
Wrist Action™ Model 75 Laboratory Shakers
Bench top shakers with swirling action of hand mixing
Have Questions?
Change view
Catalog Number
Frequency
Voltage
No. of Positions
Model
FIS14-400-123
50 Hz
220 V
8
75-BB
FIS14-400-121
50 Hz
220 V
4
75-AA
2 Options
Catalog number
FIS14-400-123
Price (SGD)
17,775.00
Each
Add to cart
Frequency:
50 Hz
Voltage:
220 V
No. of Positions:
8
Model:
75-BB
Price (SGD)
17,775.00
Each
Add to cart
Single Speed with Timer
Material: Aluminum and steel
Max tested Load at 4.5kg (10 lb.)
Color: Blue and white
Specifications
Speed Range
385 rpm
Voltage
220 V
Product Type
Laboratory Shaker
Frequency
50 Hz
Weight (English)
70 lb.
Weight (Metric)
31.7 kg
Model
75-BB
Includes
Side arm shafts, Clamps, Power cord, Operations manual, Allen Wrench, Lever arm brake
No. of Positions
8