Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Material | Packaging | Type |
---|---|---|---|---|
FIS14-387-067 | 100 mL | Polystyrene | 10 Bags of 10 | Reagent Reservoir |
FKS10495973 | 75 mL | Polypropylene | 10 pcs /bag | Reservoir Lid |
8085 | 100 mL | Polystyrene | 10 Bags of 10 | Reagent Reservoir |
FKS10141873 | 100 mL | Polystyrene | 10 Bags of 10 | Reagent Reservoir |
8086 | 100 mL | Polystyrene | 50 Individually wrapped | Reagent Reservoir |
FIS14-387-068 | 100 mL | Polystyrene | 50 Individually wrapped | Reagent Reservoir |
FKS10375023 | 100 mL | Polystyrene | 50 Individually wrapped | Reagent Reservoir |
FIS14-387-069 | 25 mL | Polystyrene | 10 Bags of 10 | Reagent Reservoir |
FKS10756354 | 25 mL | Polystyrene | 10 Bags of 10 | Reagent Reservoir |
8094 | 25 mL | Polystyrene | 50 Individually wrapped | Reagent Reservoir |
8095 | 25 mL | Polystyrene | 10 Bags of 10 | Reagent Reservoir with Divider |
FIS14-387-071 | 25 mL | Polystyrene | 10 Bags of 10 | Reagent Reservoir with Divider |
FKS10225743 | 25 mL | Polystyrene | 10 Bags of 10 | Reagent Reservoir with Divider |
FIS14-387-072 | 25 mL | Polystyrene | 50 Individually wrapped | Reagent Reservoir with Divider |
Accomodate a variety of applications with uniquely designed, innovative Thermo Scientific™ Matrix Reagent Reservoirs. These sterile reservoirs offer an extra-wide base to add rigidity and stability, helping to avoid spills. The 25mL divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizing recovery if using only a single channel and a small amount of reagent.