Search Fisher Scientific
Handle reagents on automated systems simply and economically.
| Catalog Number | Channels | Sterility | Description | Material |
|---|---|---|---|---|
| FIS14-387-034 | 96 | Sterile | Deepwell | Polypropylene |
| 1064-15-8 | 96 | Sterile | Polypropylene | |
| FIS14-387-035 | 384 | Non-sterile | Polypropylene | |
| 1064-05-6 | 96 | Non-sterile | Deepwell | Plastic |
Thermo Scientific™ Automation Reservoirs provide standard SBS dimensions ensuring compatibility with plate stackers and handlers, which enables you to easily handle reagents on automated systems economically. Reservoirs are disposable with no need for washing, allowing more reservoirs to be used per run. Built-in grooves in reservoir bottom reduce reagent waste and allow consistent aspiration.
Removable Splash Baffles: