Class A PMP Volumetric Flasks with NS Stoppers
Actual product may vary
BrandTech
Class A PMP Volumetric Flasks with NS Stoppers
These Class A Volumetric Flasks with NS Stoppers, calibrated 'to contain', are an excellent break-resistant alternative to glass in the laboratory.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Standard Taper JointTolerance
FIS14-379-37510 mL10/190.04 mL
FIS14-379-2191000 mL24/290.4 mL
Catalog number FIS14-379-375
Price (SGD)
134.00
Pack
Capacity (Metric):
10 mL
Standard Taper Joint:
10/19
Tolerance:
0.04 mL
Price (SGD)
134.00
Pack
  • Meet Class A tolerances per DIN EN ISO 1042
  • Manufactured from crystal-clear polymethylpentene
  • Supplied certificate includes mean test value and lot (lot and year imprinted on product)
  • Thermal stress up to 121°C (autoclaving) does not cause permanent exceeding of the tolerance limits
  • However, to preserve markings, cleaning at no higher than 60°C is recommended
  • Available in a range of capacities and stopper sizes
Specifications
CalibrationCalibrated 'to contain' per DIN EN ISO 1042
Closure MaterialPolypropylene
ColorTranslucent
Capacity (Metric)10 mL
MaterialPMP
Height (Metric)90 mm
IncludesImprinted lot number, batch certificate
Product TypeVolumetric Flask
Standard Taper Joint10/19
Temperature Range (Metric)121°C max.
Tolerance0.04 mL
AutoclavableAutoclavable
DisposableReusable
GraduatedNo
Mouth StyleJoint
SterilityNon-sterile
Bottom ShapeRound
Closure TypeNS Stopper
Unit SizePack