Titrette™ Digital Bottle-Top Burettes
Actual product may vary
Titrette™ Digital Bottle-Top Burettes
Titrette™ Digital Bottle-Top Burettes
BrandTech
Titrette™ Digital Bottle-Top Burettes
Provides accuracy and precision within the tolerances of Class A glass burettes. Titrette™ Digital Bottle-top Burets are suitable for most titrants in concentrations up to 1 mol/L. Ideal for general chemistry, wastewater treatment applications, food/beverage analysis, industrial titrations, environmental fieldwork, and more.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Interface
FIS14-379-34350 mLWithout Interface
FIS14-379-34225 mLWithout Interface
FKS1070316425 mLWithout Interface
FKS1070335450 mLWithout Interface
FIS14-379-34025 mLRS232 Interface
FKS1068742525 mLRS232 Interface
FIS14-379-34150 mLRS232 Interface
FKS1076916550 mLRS232 Interface
FIS13-690-10110 mLRS232 Interface
Catalog number FIS14-379-343
Price (SGD)
4,876.00
Each
Volume (Metric):
50 mL
Interface:
Without Interface
Price (SGD)
4,876.00
Each
Provides accuracy and precision within the tolerances of Class A glass burettes. BrandTech™ BRAND™ Titrette™ Digital Bottle-top Burets are suitable for most titrants in concentrations up to 1 mol/L. Ideal for general chemistry, wastewater treatment applications, food/beverage analysis, industrial titrations, environmental fieldwork, and more.
The Titrette™ bottle-top burette provides accuracy and precision within the tolerances of Class A glass burettes. Suitable for most titrants in concentrations up to 1 mol/L, it is ideal for general chemistry, wastewater treatment applications, food/beverage analysis, industrial titrations, environmental fieldwork, and more. The Titrette will enable you to perform routine titrations quickly, accurately, and with ease.
  • Smooth operation for exceptionally sensitive drop-wise titrations
  • Avoids the risk of spills from poured transfers
  • Eliminates meniscus reading errors
  • Switch automatically between filling and titrating
  • Meets tolerances for Class A glass burettes per ISO and ASTM standards
  • Optional RS-232 interface available for direct data transmission to a PC
  • Use for a variety of applications including general chemistry, water treatment, food/beverage analysis, industrial titrations, and environmental work in the lab or field
Specifications
Accuracy0.06 %
Product TypeDigital Bottletop Dispenser
Angle90°
AutoclavableNot Autoclavable
Certifications/ComplianceDIN EN ISO 385, ASTM E287
ColorGray
MaterialGlass
Volume (Metric)50 mL
DisplayDigital
For Use With (Application)Environmental Fieldwork, Food or Beverage Analysis, General Chemistry, Industrial Titrations, Wastewater Treatment Applications
Includes(2) AAA Batteries, (2) Amber Inspection Windows, (2) Colored Light Shield Inspection Windows, (3) Bottle Thread Adapters, Operating Manual, Performance Certificate, Telescopic Filling Tube
IncrementsApprox. 20 μL
InterfaceWithout Interface
Precision0.02 %
Temperature (English)59°F to 104°F
Temperature (Metric)15°C to 40°C
Viscosity RatingUp to 500 mm 2/s
Voltage1.5 V
Unit SizeEach