Measuring Scoops
Actual product may vary
BrandTech
Measuring Scoops
In assorted sizes
Have Questions?
Catalog NumberCapacity (Metric)Material
FIS14-379-2901250 mLHDPE
Catalog number FIS14-379-290
Price (SGD)
341.00
Pack
Capacity (Metric):
1250 mL
Material:
HDPE
Price (SGD)
341.00
Pack
  • Choose polypropylene or polyethylene
  • Metric volume (mL) marked on handle
Specifications
Capacity (English)42.26 oz.
Capacity (Metric)1250 mL
Product TypeMeasuring Scoop
For Use With (Application)For measuring samples
Length (English)101.6 in.
MaterialHDPE
Length (Metric)40 cm
AutoclavableNot Autoclavable
ShapeScoop
Unit SizePack