Polypropylene Graduated Pitchers
Actual product may vary
BrandTech
Polypropylene Graduated Pitchers
VITLAB™ polypropylene graduated pitchers in nesting or non-nesting styles. Non-nesting versions have molded graduations for durability, and are available with or without highly legible blue screened overprint.
Have Questions?
Catalog NumberCapacity (Metric)Graduations
FIS14-379-2775000 mLBlue Screened Molded
Catalog number FIS14-379-277
Price (SGD)
728.00
Pack
Capacity (Metric):
5000 mL
Graduations:
Blue Screened Molded
Price (SGD)
728.00
Pack
  • Polypropylene pitchers are lighter than glass for easy handling
  • Good chemical compatibility
  • High translucence for easy verification of volume and contents
Specifications
Capacity169.07 oz., 5000 mL
Capacity (Metric)5000 mL
Capacity (English)169.07 oz.
ColorTranslucent
Diameter (English)8.26 in.
MaterialPolypropylene
Quantity6/Pk.
SterilityNon-sterile
GraduationsBlue Screened Molded
Closure IncludedNo
DisposableReusable
GraduatedYes
ShapeRound
Height (Metric)270 mm
Product TypePouring Pitcher
Width (English)8.0 in.
Height (English)10.62 in.
Diameter (Metric)210 mm