Class A Certified Graduated Cylinders
Actual product may vary
BrandTech
Class A Certified Graduated Cylinders
VITLAB polymethylpentene graduated cylinders, certified Class A to DIN 12681 and ISO 6706.
Have Questions?
Catalog NumberCapacity (Metric)Graduations
FIS14-379-14925 mL0.5 mL
Catalog number FIS14-379-149
Price (SGD)
73.00
Pack
Capacity (Metric):
25 mL
Graduations:
0.5 mL
Price (SGD)
73.00
Pack
  • Molded graduations with ring marks at major increments
  • Imprinted lot and year
  • Supplied certificate includes mean test value and lot
Specifications
Capacity (Metric)25 mL
MaterialPMP
Tolerance±0.25 mL
Certifications/ComplianceDIN 12681, ISO 6706
ClassClass A
CalibrationTo Contain
Diameter (English)0.82 in.
Diameter (Metric)21 mm
Graduations0.5 mL
AutoclavableAutoclavable
ShapeRound
Base TypeHexagonal
Height (English)6.65 in.
Height (Metric)170 mm
Product TypeGraduated Cylinder
Unit SizePack