Safetex™ No Touch™ Polypropylene Slide Holders
Safetex™ No Touch™ Polypropylene Slide Holders
Actual product may vary
Fisherbrand
Safetex™ No Touch™ Polypropylene Slide Holders
Patented window design allows for easy patient identification and helps ensure that slides reach the lab intact and unbroken
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
FIS14-378-682 Slides
FIS14-372-201 Slide
Catalog number FIS14-378-68
Price (SGD)
-
Quantity:
2 Slides
  • Safetex plastic
  • Available in one- and two-slide configurations
  • Sturdy polypropylene
  • No Touch security prevents the specimen from touching the slide holder cover
  • Holds one or two 3 x 1 in. slides
Specifications
MaterialPolypropylene
Quantity2 Slides
Unit SizeCase