Plastic Spectrophotometry Cuvettes
Actual product may vary
BrandTech
Plastic Spectrophotometry Cuvettes
BRAND™ Plastic Spectrophotometry Cuvettes with mold-cavity matched for the lowest variability in extinction coefficient.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)MaterialQuantity
FIS14-377-0173 mLPMMA100 Pk.
FIS14-377-0154.5 mLPMMA100 Pk.
FIS14-377-0123 mLPolystyrene500 Pk.
Catalog number FIS14-377-017
Price (SGD)
91.00
Pack
Capacity (Metric):
3 mL
Material:
PMMA
Quantity:
100 Pk.
Price (SGD)
91.00
Pack
BRAND spectrophotometry cuvettes. Mold-cavity matched for the lowest variability in extinction coefficient.
  • Mold cavity sorted spectrophotometry cuvettes with 2 clear windows
Specifications
SterilityNon-sterile
TypeSpectrophotometer Cuvette
DescriptionSemi-micro
ColorClear
DisposableSingle-use
Capacity (Metric)3 mL
MaterialPMMA
IncludesArrow mark to indicate direction of transmission and reduce variation
Certifications/ComplianceLife Science Quality
Quantity100 Pk.
Path Length10 mm
Wavelength Range300 nm
Height (Metric)45 mm
For Use With (Equipment)Spectrophotometers, Photometers
Width (Metric)12.5 mm
For Use With (Application)Spectrophotometry
Lightpath10 mm
ShapeRectangle
Unit SizePack