▸
Tubes
▸
Auto Analyzer Tubes
▸
False Bottom Analyzer Tube
Actual product may vary
False Bottom Analyzer Tube
Designed for high-volume analyzer aspiration probes incapable of reaching the bottoms of larger sample tubes
Have Questions?
Change view
Catalog Number
Color
Product Type
Material
FIS14-375-441
White
Screw Cap
Plastic
FIS14-375-439
Clear
False Bottom Analyzer Vial
Polypropylene
2 Options
Catalog number
FIS14-375-441
Price (SGD)
475.00
Case
Add to cart
Color:
White
Product Type:
Screw Cap
Material:
Plastic
Price (SGD)
475.00
Case
Add to cart
Ideal for primary or secondary sampling as well as storage
No need for micro sample cups and liquid transfer devices
Accept bar codes and fit into most high volume analyzer racks
Compatible with Hitachi™, Olympus™, Corning™, Ortho (Kodak™), VITROS™ (750 and 250 series), and Beckman™ clinical chemistry analyzer
Specifications
Closure Type
Screw
For Use With (Equipment)
Starplex false bottom auto analyzer tube
Sterility
Non-sterile
Color
White
Material
Plastic
Product Type
Screw Cap