Disposable Sterile Scoops
Actual product may vary
Disposable Sterile Scoops
Curved handle for easier pickup
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Length (Metric)
FIS14-375-251125 mL19 cm
FIS14-375-25060 mL19 cm
FIS14-375-252250 mL20 cm
Catalog number FIS14-375-251
Price (SGD)
-
Capacity (Metric):
125 mL
Length (Metric):
19 cm
Curved handle for easier pickup
  • Contaminant-free
  • Flat-top design ensures accuracy to ±2% of capacity
  • Flat bottom keeps scoop upright
  • Polystyrene construction
Specifications
SterilitySterile
ShapeFlat top
Capacity (English)4 oz.
Capacity (Metric)125 mL
DisposableSingle-use
Product TypeScoop
Length (English)7.5 in.
MaterialPolystyrene
Handle TypeCurved
Length (Metric)19 cm
Unit SizeCase