Spoonula™ Lab Spoon
Actual product may vary
Fisherbrand
Spoonula™ Lab Spoon
Fisherbrand Spoonula™ Lab Spoon is used for transferring crystalline materials, crushing lumps, scraping, or even chopping!
Have Questions?
Catalog NumberLength (Metric)
FIS14-375-1022.86 cm
Catalog number FIS14-375-10
Price (SGD)
84.00
Pack
Length (Metric):
22.86 cm
Price (SGD)
84.00
Pack
Fisherbrand Spoonula™ Lab Spoon is used for transferring crystalline materials, crushing lumps, scraping, or even chopping!
Fisherbrand™ Spoonula™ Lab Spoon is used for transferring crystalline materials, crushing lumps, scraping, or even chopping. Our spatulas are designed to exceed performance expectations -This is the Fisherbrand Advantage. Available in variety of lengths.
  • Made from ASTM™ standard stainless-steel construction
  • Polished finishing
  • Spoon end: 1.25L x 0.56 in.W (32 x 14mm)
  • Spatula end: 2L x 0.31 in.W (51 x 7.9mm)
Specifications
ShapeSpoon End, Spatula End
Certifications/ComplianceASTM
Product TypeSpoonula™ Lab Spoon
FinishPolished
For Use With (Application)Designed for transferring crystalline materials, crushing lumps, scraping or chopping
Length (English)9 in.
IncludesDouble ended with 1.25 L x 0.56 in. W spoon end and 2 L x 0.31 in. W spatula end on other
MaterialStainless Steel
Length (Metric)22.86 cm
Unit SizePack